Netherlands
Order Entry
Biohazardous Plastic Waste Can, Foot-Operated
Biohazardous Plastic Waste Can, Foot-Operated
New Product
Certificates
Biohazardous Plastic Waste Can, Foot-Operated
New Product
Product Information
  • Catalog #
    Supplier #
    Material
    Capacity
    Colour
    Type
    W×D×H
    Availability
    Price
  • JUSU943BIO
    943BIO
    HDPE
    6 gallons (22,7 L)
    Red
    Foot-operated, self-closing
    40,64×34,29×40,64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
    Supplier Article Number: 943BIO
    Local Article Number: JUSU943BIO
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • JUSU945BIO
    945BIO
    HDPE
    10 gallons (37,85 L)
    Red
    Foot-operated, self-closing
    55,88×45,72×45,72 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 153,00 €
    153,00 €
    1 * 1 ST
       153,00 €
    Supplier Article Number: 945BIO
    Local Article Number: JUSU945BIO
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • JUSU947BIO
    947BIO
    HDPE
    14 gallons (53 L)
    Red
    Foot-operated, self-closing
    55,88×45,72×53,34 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
    Supplier Article Number: 947BIO
    Local Article Number: JUSU947BIO
    Specifications:
    New Product
    Specifications
    Environmentally Preferable
    New Product